Anti-AAK1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB127614-100 ORB127614-200
BIRBORB127614-100EA 0 EUR
BIRBORB127614-100 BIRBORB127614-200
Anti-AAK1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: AAK1 (AP2 associated kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR