Anti-APPBP2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB127691-100 ORB127691-200
BIRBORB127691-100EA 0 EUR
BIRBORB127691-100 BIRBORB127691-200
Anti-APPBP2 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: APPBP2 (amyloid beta precursor protein (cytoplasmic tail) binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR