Anti-CORO1A Rabbit Polyclonal Antibody
Proveedor: Biorbyt
ORB127915-100 ORB127915-200
BIRBORB127915-100EA 0 EUR
BIRBORB127915-100 BIRBORB127915-200
Anti-CORO1A Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CORO1A (coronin, actin binding protein, 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat