Anti-EIF2B4 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB128013-100 ORB128013-200
BIRBORB128013-100EA 0 EUR
BIRBORB128013-100 BIRBORB128013-200
Anti-EIF2B4 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: EIF2B4 (eukaryotic translation initiation factor 2B, subunit 4 delta, 67kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR