Anti-PRKCG Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB128579-100 ORB128579-200
BIRBORB128579-100EA 0 EUR
BIRBORB128579-100 BIRBORB128579-200
Anti-PRKCG Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PRKCG (protein kinase C, gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR