Anti-PSMD6 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB128593-100 ORB128593-200
BIRBORB128593-100EA 0 EUR
BIRBORB128593-100 BIRBORB128593-200
Anti-PSMD6 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PSMD6 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR