Anti-TNNI3K Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB128856-100 ORB128856-200
BIRBORB128856-100EA 0 EUR
BIRBORB128856-100 BIRBORB128856-200
Anti-TNNI3K Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: TNNI3K (TNNI3 interacting kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR