Anti-EPHA2/3/4 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB128954-100 ORB128954-200
BIRBORB128954-100EA 0 EUR
BIRBORB128954-100 BIRBORB128954-200
Anti-EPHA2/3/4 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: EPHA2/3/4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR