Anti-PRKCA Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB129720-100 ORB129720-200
BIRBORB129720-100EA 0 EUR
BIRBORB129720-100 BIRBORB129720-200
Anti-PRKCA Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PRKCA (protein kinase C, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR