Anti-RAC1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB129728-100 ORB129728-200
BIRBORB129728-100EA 0 EUR
BIRBORB129728-100 BIRBORB129728-200
Anti-RAC1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: RAC1 (ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR