Anti-NFASC Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB136801-100 ORB136801-200 ORB136801-50
BIRBORB136801-100EA 0 EUR
BIRBORB136801-100 BIRBORB136801-200 BIRBORB136801-50
Anti-NFASC Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: NFASC (neurofascin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR