Anti-CROT Rabbit polyclonal antibody unconjugated

Proveedor: Biorbyt
ORB99356-100 ORB99356-20
BIRBORB99356-100EA 0 EUR
BIRBORB99356-100 BIRBORB99356-20
Anti-CROT Rabbit polyclonal antibody unconjugated
Anticuerpos
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: CROT (carnitine O-octanoyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR