Anti-TGF beta 3 Rabbit Polyclonal Antibody

Proveedor: Abcam
AB15537-500 AB15537-100 AB15537-1
ABCAAB15537-500EA 975 EUR
ABCAAB15537-500 ABCAAB15537-100 ABCAAB15537-1
Anti-TGF beta 3 Rabbit Polyclonal Antibody
Anticuerpos
Rabbit polyclonal to TGF beta 3.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: TGF beta 3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Rabbit;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR