Anti-PKC beta 2 Rabbit Monoclonal Antibody [clone: Y125]
Proveedor: Abcam
AB32026-100 AB32026-1ML
ABCAAB32026-100EA 819 EUR
ABCAAB32026-100 ABCAAB32026-1ML
Anti-PKC beta 2 Rabbit Monoclonal Antibody [clone: Y125]
Anticuerpos
Rabbit monoclonal [Y125] to PKC beta 2.
Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: PKC beta 2
Clonality: Monoclonal
Clone: Y125
Conjugation:
Epitope: ab32026 reacts with an epitope located in the region near the C terminus of PKC beta 2.
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human