Anti-CD3 epsilon Rabbit Monoclonal Antibody [clone: E272]

Proveedor: Abcam
AB32186-100 AB32186-1ML
ABCAAB32186-100EA 475 EUR
ABCAAB32186-100 ABCAAB32186-1ML
Anti-CD3 epsilon Rabbit Monoclonal Antibody [clone: E272]
Anticuerpos
Rabbit monoclonal [E272] to CD3 epsilon.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD3 epsilon
Clonality: Monoclonal
Clone: E272
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR