Anti-MAP1LC3A Rabbit Monoclonal Antibody [clone: EP1983Y]
Proveedor: Abcam
AB52768-100 AB52768-1ML AB52768-20UL
ABCAAB52768-100EA 475 EUR
ABCAAB52768-100 ABCAAB52768-1ML ABCAAB52768-20UL
Anti-MAP1LC3A Rabbit Monoclonal Antibody [clone: EP1983Y]
Anticuerpos
Rabbit monoclonal [EP1983Y] to MAP1LC3A - Autophagosome Marker.
Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: MAP1LC3A
Clonality: Monoclonal
Clone: EP1983Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat;Human