Anti-beta III Tubulin Rabbit Monoclonal Antibody [clone: EPR1568Y]
Proveedor: Abcam
AB68193-100 AB68193-1ML
ABCAAB68193-100EA 688 EUR
ABCAAB68193-100 ABCAAB68193-1ML
Anti-beta III Tubulin Rabbit Monoclonal Antibody [clone: EPR1568Y]
Anticuerpos
Rabbit monoclonal [EPR1568Y] to beta III Tubulin.
Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: beta III Tubulin
Clonality: Monoclonal
Clone: EPR1568Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat