Anti-beta 2 Microglobulin Rabbit Monoclonal Antibody [clone: EP2978Y]

Proveedor: Abcam
AB75853-100 AB75853-1ML
ABCAAB75853-100EA 788 EUR
ABCAAB75853-100 ABCAAB75853-1ML
Anti-beta 2 Microglobulin Rabbit Monoclonal Antibody [clone: EP2978Y]
Anticuerpos
Rabbit monoclonal [EP2978Y] to beta 2 Microglobulin.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: beta 2 Microglobulin
Clonality: Monoclonal
Clone: EP2978Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR