Anti-Cytokeratin 16/K16 Rabbit Monoclonal Antibody [clone: EP1615Y]

Proveedor: Abcam
AB76416-100 AB76416-1ML
ABCAAB76416-100EA 744 EUR
ABCAAB76416-100 ABCAAB76416-1ML
Anti-Cytokeratin 16/K16 Rabbit Monoclonal Antibody [clone: EP1615Y]
Anticuerpos
Rabbit monoclonal [EP1615Y] to Cytokeratin 16/K16.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Cytokeratin 16/K16
Clonality: Monoclonal
Clone: EP1615Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR