Anti-Integrin alpha 4/CD49D Rabbit Monoclonal Antibody [clone: EPR1355Y]

Proveedor: Abcam
AB81280-100 AB81280-1ML
ABCAAB81280-100EA 682 EUR
ABCAAB81280-100 ABCAAB81280-1ML
Anti-Integrin alpha 4/CD49D Rabbit Monoclonal Antibody [clone: EPR1355Y]
Anticuerpos
Rabbit monoclonal [EPR1355Y] to Integrin alpha 4/CD49D.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Integrin alpha 4/CD49D
Clonality: Monoclonal
Clone: EPR1355Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR