Anti-SCP3 Mouse Monoclonal Antibody [clone: Cor 10G11/7]
Proveedor: Abcam
AB97672-100 AB97672-1MG
ABCAAB97672-100EA 657 EUR
ABCAAB97672-100 ABCAAB97672-1MG
Anti-SCP3 Mouse Monoclonal Antibody [clone: Cor 10G11/7]
Anticuerpos
Mouse monoclonal [Cor 10G11/7] to SCP3.
Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SCP3
Clonality: Monoclonal
Clone: Cor 10G11/7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Rat