Anti-alpha Actinin 4 Rabbit Monoclonal Antibody [clone: EPR4410]

Proveedor: Abcam
AB108201-100 AB108201-1ML
ABCAAB108201-100EA 475 EUR
ABCAAB108201-100 ABCAAB108201-1ML
Anti-alpha Actinin 4 Rabbit Monoclonal Antibody [clone: EPR4410]
Anticuerpos
Rabbit monoclonal [EPR4410] to alpha Actinin 4.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: alpha Actinin 4
Clonality: Monoclonal
Clone: EPR4410
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR