Anti-Transferrin Receptor Rabbit Monoclonal Antibody [clone: EPR4012]

Proveedor: Abcam
AB108985-100 AB108985-1ML
ABCAAB108985-100EA 707 EUR
ABCAAB108985-100 ABCAAB108985-1ML
Anti-Transferrin Receptor Rabbit Monoclonal Antibody [clone: EPR4012]
Anticuerpos
Rabbit monoclonal [EPR4012] to Transferrin Receptor.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Transferrin Receptor
Clonality: Monoclonal
Clone: EPR4012
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR