Anti-PSMD14 Rabbit Monoclonal Antibody [clone: EPR4257]

Proveedor: Abcam
AB109123-100 AB109123-1ML
ABCAAB109123-100EA 725 EUR
ABCAAB109123-100 ABCAAB109123-1ML
Anti-PSMD14 Rabbit Monoclonal Antibody [clone: EPR4257]
Anticuerpos
Rabbit monoclonal [EPR4257] to PSMD14.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PSMD14
Clonality: Monoclonal
Clone: EPR4257
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR