Anti-PAX6 Rabbit Monoclonal Antibody [clone: EPR3352(2)]

Proveedor: Abcam
AB109233-100 AB109233-20UL AB109233-1ML
ABCAAB109233-100EA 694 EUR
ABCAAB109233-100 ABCAAB109233-20UL ABCAAB109233-1ML
Anti-PAX6 Rabbit Monoclonal Antibody [clone: EPR3352(2)]
Anticuerpos
Rabbit monoclonal [EPR3352(2)] to PAX6.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PAX6
Clonality: Monoclonal
Clone: EPR3352(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR