Anti-PSMA3 Rabbit Monoclonal Antibody [clone: EPR5455]

Proveedor: Abcam
AB109532-100 AB109532-10 AB109532-1ML
ABCAAB109532-100EA 682 EUR
ABCAAB109532-100 ABCAAB109532-10 ABCAAB109532-1ML
Anti-PSMA3 Rabbit Monoclonal Antibody [clone: EPR5455]
Anticuerpos
Rabbit monoclonal [EPR5455] to PSMA3.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PSMA3
Clonality: Monoclonal
Clone: EPR5455
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR