Anti-VEGFB Rabbit Monoclonal Antibody [clone: EPR4555]

Proveedor: Abcam
AB110649-100 AB110649-1ML
ABCAAB110649-100EA 669 EUR
ABCAAB110649-100 ABCAAB110649-1ML
Anti-VEGFB Rabbit Monoclonal Antibody [clone: EPR4555]
Anticuerpos
Type: Primary
Antigen: VEGFB (vascular endothelial growth factor B)
Clonality: monoclonal
Clone: EPR4555
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR