Anti-FRDA Mouse Monoclonal Antibody [clone: 17A11]
Proveedor: Abcam
AB113691-100 AB113691-1
ABCAAB113691-100EA 563 EUR
ABCAAB113691-100 ABCAAB113691-1
Anti-FRDA Mouse Monoclonal Antibody [clone: 17A11]
Anticuerpos
Mouse monoclonal [17A11] to Frataxin.
Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: FRDA
Clonality: Monoclonal
Clone: 17A11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Rat;Bovine;Human