Anti-alpha 1 Sodium Potassium ATPase (phospho Y10) Rabbit Monoclonal Antibody [clone: EPR2375(2)]

Proveedor: Abcam
AB124677-100 AB124677-1ML
ABCAAB124677-100EA 475 EUR
ABCAAB124677-100 ABCAAB124677-1ML
Anti-alpha 1 Sodium Potassium ATPase (phospho Y10) Rabbit Monoclonal Antibody [clone: EPR2375(2)]
Anticuerpos
Rabbit monoclonal [EPR2375(2)] to alpha 1 Sodium Potassium ATPase (phospho Y10).

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: alpha 1 Sodium Potassium ATPase
Clonality: Monoclonal
Clone: EPR2375(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR