Anti-IL-7 Rabbit Monoclonal Antibody [clone: EPR6266]

Proveedor: Abcam
AB125029-100 AB125029-1ML
ABCAAB125029-100EA 700 EUR
ABCAAB125029-100 ABCAAB125029-1ML
Anti-IL-7 Rabbit Monoclonal Antibody [clone: EPR6266]
Anticuerpos
Rabbit monoclonal [EPR6266] to IL-7.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: IL-7
Clonality: Monoclonal
Clone: EPR6266
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR