Anti-IL-28A+IL-28B Rabbit Monoclonal Antibody [clone: EPR5328(2)]

Proveedor: Abcam
AB126713-100 AB126713-1ML
ABCAAB126713-100EA 675 EUR
ABCAAB126713-100 ABCAAB126713-1ML
Anti-IL-28A+IL-28B Rabbit Monoclonal Antibody [clone: EPR5328(2)]
Anticuerpos
Rabbit monoclonal [EPR5328(2)] to IL-28A + IL-28B.

Precaución: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IL-28A+IL-28B
Clonality: Monoclonal
Clone: EPR5328(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR