Anti-HNF-4-alpha Rabbit Monoclonal Antibody [clone: EPR3649]

Proveedor: Abcam
AB133343-100 AB133343-1ML
ABCAAB133343-100EA 719 EUR
ABCAAB133343-100 ABCAAB133343-1ML
Anti-HNF-4-alpha Rabbit Monoclonal Antibody [clone: EPR3649]
Anticuerpos
Rabbit monoclonal [EPR3649] to HNF-4-alpha.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: HNF-4-alpha
Clonality: Monoclonal
Clone: EPR3649
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR