Anti-Mu Opioid Receptor Rabbit Monoclonal Antibody [clone: UMB3]

Proveedor: Abcam
AB134054-100 AB134054-1ML
ABCAAB134054-100EA 725 EUR
ABCAAB134054-100 ABCAAB134054-1ML
Anti-Mu Opioid Receptor Rabbit Monoclonal Antibody [clone: UMB3]
Anticuerpos
Rabbit monoclonal [UMB3] to Mu Opioid Receptor - C-terminal.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Mu Opioid Receptor
Clonality: Monoclonal
Clone: UMB3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR