Anti-Integrin beta 1 Rabbit Monoclonal Antibody [clone: EPR1040Y]

Proveedor: Abcam
AB134179-100 AB134179-1ML
ABCAAB134179-100EA 769 EUR
ABCAAB134179-100 ABCAAB134179-1ML
Anti-Integrin beta 1 Rabbit Monoclonal Antibody [clone: EPR1040Y]
Anticuerpos
Rabbit monoclonal [EPR1040Y] to Integrin beta 1.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Integrin beta 1
Clonality: Monoclonal
Clone: EPR1040Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR