Anti-FSH beta Rabbit Monoclonal Antibody [clone: EPR7930]

Proveedor: Abcam
AB150425-100 AB150425-1ML
ABCAAB150425-100EA 688 EUR
ABCAAB150425-100 ABCAAB150425-1ML
Anti-FSH beta Rabbit Monoclonal Antibody [clone: EPR7930]
Anticuerpos
Rabbit monoclonal [EPR7930] to FSH beta.

Precaución: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: FSH beta
Clonality: Monoclonal
Clone: EPR7930
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR