Anti-IgG Goat Polyclonal Antibody (CF™ 647)
Proveedor: Biotium
20042 20042-1
20042.EA 229 EUR
20042. 20042-1
Anti-IgG Goat Polyclonal Antibody (CF™ 647)
Anticuerpos
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with CF™647 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Far-red fluorescent CF™647 (Ex/Em 650/665 nm) is brighter than Cy®5 and an excellent alternative to Alexa Fluor® 647 for antibody conjugates.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF647
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse