Anti-IgG Donkey Polyclonal Antibody (CF™ 633)

Proveedor: Biotium
20124 20124-1
BTIU20124EA 359 EUR
BTIU20124 20124-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 633)
Anticuerpos
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Mouse
Order Now

ESPECIFICACIONES

Simbolo antígeno IgG
Environmentally Preferable
Tipo de anticuerpo Secundario
Clonalidad Polyclonal
Conjugado CF® 633
Reactividad Mouse
Huésped Donkey
Isotipo
Western blot Yes
ELISA Yes
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Citometría de flujo Yes
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR