Anti-IgG Goat Polyclonal Antibody (CF™ 350)

Proveedor: Biotium
20140 20140-1
20140.EA 300 EUR
20140. 20140-1
Anti-IgG Goat Polyclonal Antibody (CF™ 350)
Anticuerpos
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 350 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR