Anti-IgG Goat Polyclonal Antibody (Biotin)

Proveedor: Biotium
20183 20183-1
20183.EA 300 EUR
20183. 20183-1
Anti-IgG Goat Polyclonal Antibody (Biotin)
Anticuerpos
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

ESPECIFICACIONES

Simbolo antígeno IgG
Environmentally Preferable
Tipo de anticuerpo Secundario
Clonalidad Polyclonal
Conjugado Biotin
Reactividad Mouse
Huésped Goat
Isotipo
Western blot Yes
ELISA Yes
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Citometría de flujo Yes
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR