Anti-IgG Donkey Polyclonal Antibody (CF™ 680R)

Proveedor: Biotium
20194 20194-1
20194.EA 368 EUR
20194. 20194-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 680R)
Anticuerpos
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 680R Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR