Anti-Fluorescein Mouse Monoclonal Antibody (FITC (Fluorescein))

Proveedor: Biotium
20212 20212-1
20212.EA 385 EUR
20212. 20212-1
Anti-Fluorescein Mouse Monoclonal Antibody (FITC (Fluorescein))
Anticuerpos
Type: Tag
Antigen: Fluorescein (fluorescein)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR