Anti-IgG Chicken Polyclonal Antibody (CF™ 594)

Proveedor: Biotium
20221 20221-1
20221.EA 300 EUR
20221. 20221-1
Anti-IgG Chicken Polyclonal Antibody (CF™ 594)
Anticuerpos
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Chicken
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR