Anti-6X His tag Mouse Monoclonal Antibody (CF®594)

Proveedor: Biotium

20229.EA 544 EUR
20229.
Anti-6X His tag Mouse Monoclonal Antibody (CF®594)
Anticuerpos
Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: monoclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR