Anti-IgG2a Goat Polyclonal Antibody (CF™ 594)
Proveedor: Biotium
20259 20259-1
20259.EA 385 EUR
20259. 20259-1
Anti-IgG2a Goat Polyclonal Antibody (CF™ 594)
Anticuerpos
Type: Secondary
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: CF® 594 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse