Anti-IgG Rabbit Polyclonal Antibody (CF™ 543)

Proveedor: Biotium
20307 20307-1
20307.EA 359 EUR
20307. 20307-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 543)
Anticuerpos
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 543 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR