Anti-NKX3-1 Mouse Monoclonal Antibody [clone: 0361]
Proveedor: Novus Biologicals
NB100-1828 NB100-1828SS
NOVUNB100-1828EA 702 EUR
NOVUNB100-1828 NOVUNB100-1828SS
Anti-NKX3-1 Mouse Monoclonal Antibody [clone: 0361]
Anticuerpos
Type: Primary
Antigen: NKX3-1 (NK3 homeobox 1)
Clonality: monoclonal
Clone: 361
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Mouse, Rat