Anti-SSEA1 Mouse Monoclonal Antibody [clone: MC-480]

Proveedor: Novus Biologicals
NB100-1831 NB100-1831SS
NOVUNB100-1831EA 606 EUR
NOVUNB100-1831 NOVUNB100-1831SS
Anti-SSEA1 Mouse Monoclonal Antibody [clone: MC-480]
Anticuerpos
Type: Primary
Antigen: SSEA1
Clonality: monoclonal
Clone: MC-480
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR