Anti-HP1 alpha Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB100-2518 NB100-2518SS
NOVUNB100-2518EA 598 EUR
NOVUNB100-2518 NOVUNB100-2518SS
Anti-HP1 alpha Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: HP1 alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR