Anti-SIRT6 Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB100-2523 NB100-2523SS
NOVUNB100-2523EA 591 EUR
NOVUNB100-2523 NOVUNB100-2523SS
Anti-SIRT6 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SIRT6 (sirtuin 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR