Anti-HIF1A Rabbit Polyclonal Antibody

Proveedor: Novus Biologicals
NB100-654 NB100-654SS
NOVUNB100-654EA 623 EUR
NOVUNB100-654 NOVUNB100-654SS
Anti-HIF1A Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR